siRNA Details
Gene Symbol: LOC93429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027003.1 | 6 | 1608 | |||
GenBank mRNA | AL137376.1 | - | 6 | 1607 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC93429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027003.1 | 6 | 1608 | |||
GenBank mRNA | AL137376.1 | - | 6 | 1607 |