siRNA Details
Gene Symbol: RAET1E-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045126.1 | 1 | 216 | |||
GenBank mRNA | BC016788.1 | - | 1 | 220 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RAET1E-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045126.1 | 1 | 216 | |||
GenBank mRNA | BC016788.1 | - | 1 | 220 |