siRNA Details
Gene Symbol: SCGB1B2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027620.3 | 4 | 1817 | |||
GenBank mRNA | CD173361.1 | - | 3 | 407 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SCGB1B2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027620.3 | 4 | 1817 | |||
GenBank mRNA | CD173361.1 | - | 3 | 407 |