siRNA Details
Gene Symbol: RAB11B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038237.1 | 1 | 645 | |||
GenBank mRNA | BC014506.1 | - | 1 | 639 | ||
BG031227.1 | - | 1 | 645 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RAB11B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038237.1 | 1 | 645 | |||
GenBank mRNA | BC014506.1 | - | 1 | 639 | ||
BG031227.1 | - | 1 | 645 |