siRNA Details
Gene Symbol: RAB11B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038237.1 | 3 | 953 | |||
GenBank mRNA | BC014506.1 | - | 3 | 947 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI760955.1 | - | 3 | 646 | Chr. 19 - 8374372 - 8390384 [-] on Build GRCh38 |
