siRNA Details
Gene Symbol: LOC101927950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110842.1 | 2 | 169 | |||
GenBank mRNA | BC080548.1 | - | 2 | 169 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ674867.1 | - | 2 | 168 | Chr. 6 - 5664973 - 5695271 [-] on Build GRCh38 |
