siRNA Details
Gene Symbol: HMGN3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040671.1 | 2 | 1200 | |||
GenBank mRNA | BC042096.1 | - | 2 | 1157 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK095853.1 | - | 2 | 1284 | Chr. 6 - 79233700 - 79236080 [+] on Build GRCh38 |
