siRNA Details
Gene Symbol: TMCC1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037893.1 | 3 | 3308 | |||
GenBank mRNA | AK127796.1 | - | 3 | 3311 | ||
AL833051.1 | - | 1 | 1715 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TMCC1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037893.1 | 3 | 3308 | |||
GenBank mRNA | AK127796.1 | - | 3 | 3311 | ||
AL833051.1 | - | 1 | 1715 |