siRNA Details
Gene Symbol: LINC00904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046222.1 | 3 | 442 | |||
GenBank mRNA | BX091932.1 | - | 3 | 442 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046222.1 | 3 | 442 | |||
GenBank mRNA | BX091932.1 | - | 3 | 442 |