siRNA Details
Gene Symbol: CASC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015410.1 | 5 | 717 | |||
GenBank mRNA | AK022865.1 | - | 4 | 343 | ||
AK026189.1 | - | 5 | 717 | |||
AK096633.1 | - | 3 | 316 | |||
AK300802.1 | - | 6 | 872 | |||
AK310212.1 | - | 7 | 1021 | |||
BC044235.1 | - | 2 | 166 | |||
KP981381.1 | - | 2 | 231 |
