siRNA Details
Gene Symbol: LOC100506286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135594.1 | 2 | 1460 | |||
GenBank mRNA | AK056547.1 | - | 2 | 1459 | ||
BC014493.1 | - | 2 | 1438 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100506286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135594.1 | 2 | 1460 | |||
GenBank mRNA | AK056547.1 | - | 2 | 1459 | ||
BC014493.1 | - | 2 | 1438 |