siRNA Details
Gene Symbol: LOC101929497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120580.1 | 3 | 398 | |||
GenBank mRNA | BC030092.2 | - | 3 | 401 | ||
DB487307.1 | - | 3 | 398 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120580.1 | 3 | 398 | |||
GenBank mRNA | BC030092.2 | - | 3 | 401 | ||
DB487307.1 | - | 3 | 398 |