siRNA Details
Gene Symbol: LOC101929497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120580.1 | 3 | 461 | |||
GenBank mRNA | BC030092.2 | - | 3 | 464 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG706864.1 | - | 3 | 469 | Chr. 11 - 127271063 - 127337035 [+] on Build GRCh38 |
