siRNA Details
Gene Symbol: LOC101928504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135252.1 | 2 | 932 | |||
GenBank mRNA | BC040628.1 | - | 2 | 932 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR617315.1 | - | 1 | 187 | Chr. 14 - 81222076 - 81223409 [+] on Build GRCh38 |
