siRNA Details
Gene Symbol: LINC01138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027468.3 | 3 | 459 | |||
NR_104014.2 | 2 | 338 | ||||
GenBank mRNA | AK096104.1 | - | 1 | 58 | ||
AK309393.1 | - | 2 | 276 | |||
DA386627.1 | - | 2 | 338 |
