siRNA Details
Gene Symbol: LINC00226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033375.2 | 2 | 512 | |||
GenBank mRNA | AW205017.1 | - | 2 | 510 | ||
AW339903.1 | - | 2 | 475 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AL833490.1 | - | 4052 | Chr. 20 - 14611073 - 14615051 [+] on Build GRCh38 |
