siRNA Details
Gene Symbol: FLJ36000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027084.1 | 2 | 3388 | |||
GenBank mRNA | AK093319.1 | - | 1 | 2325 | ||
BC039120.1 | - | 2 | 3390 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ36000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027084.1 | 2 | 3388 | |||
GenBank mRNA | AK093319.1 | - | 1 | 2325 | ||
BC039120.1 | - | 2 | 3390 |