siRNA Details
Gene Symbol: LOC101927697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109888.1 | 7 | 2786 | |||
GenBank mRNA | BC047601.1 | - | 7 | 2786 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109888.1 | 7 | 2786 | |||
GenBank mRNA | BC047601.1 | - | 7 | 2786 |