siRNA Details
Gene Symbol: LOC101927815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125423.1 | 5 | 1546 | |||
GenBank mRNA | BC030605.2 | - | 5 | 1546 | ||
BC045738.1 | - | 5 | 1546 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125423.1 | 5 | 1546 | |||
GenBank mRNA | BC030605.2 | - | 5 | 1546 | ||
BC045738.1 | - | 5 | 1546 |