siRNA Details
Gene Symbol: VWA8-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039974.1 | 3, 4 | 1304 | |||
GenBank mRNA | AK092199.1 | - | 3, 4 | 1304 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: VWA8-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039974.1 | 3, 4 | 1304 | |||
GenBank mRNA | AK092199.1 | - | 3, 4 | 1304 |