siRNA Details
Gene Symbol: LOC101928238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134920.1 | 2 | 811 | |||
NR_134921.1 | 2 | 541 | ||||
GenBank mRNA | AF075036.1 | - | 2 | 474 | ||
BC042816.1 | - | 2 | 738 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134920.1 | 2 | 811 | |||
NR_134921.1 | 2 | 541 | ||||
GenBank mRNA | AF075036.1 | - | 2 | 474 | ||
BC042816.1 | - | 2 | 738 |