siRNA Details
Gene Symbol: LINC00624 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038423.2 | 2 | 509 | |||
GenBank mRNA | BC036212.1 | - | 2 | 509 | ||
BC048126.1 | - | 2 | 509 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00624 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038423.2 | 2 | 509 | |||
GenBank mRNA | BC036212.1 | - | 2 | 509 | ||
BC048126.1 | - | 2 | 509 |