siRNA Details
Gene Symbol: LOC101926951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134946.1 | 2 | 615 | |||
GenBank mRNA | BC038756.1 | - | 2 | 615 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI462451.1 | - | 2 | 619 | Chr. 13 - 61424720 - 61427946 [-] on Build GRCh38 |
