siRNA Details
Gene Symbol: CTBP1-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104331.1 | 6 | 3205 | |||
GenBank mRNA | AK092548.1 | - | 6 | 3205 | ||
AX747592.1 | - | 6 | 3205 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CTBP1-AS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104331.1 | 6 | 3205 | |||
GenBank mRNA | AK092548.1 | - | 6 | 3205 | ||
AX747592.1 | - | 6 | 3205 |