siRNA Details
Gene Symbol: ZNF638-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC044944.1 | - | 2 | 1462 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD653923.1 | - | 1 | 439 | Chr. 2 - 71375743 - 71376484 [+] on Build GRCh38 |
