siRNA Details
Gene Symbol: 4930512B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033573.1 | 1 | 72 | |||
GenBank mRNA | AK015558.1 | - | 1 | 82 | ||
AK015769.1 | - | 1 | 129 | |||
AK029702.1 | - | 1 | 61 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715108.1 | - | 1 | 82 | Chr. 12 - 69790335 - 69840477 [+] on Build GRCm38 |
