siRNA Details
Gene Symbol: Tmem134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI789565.1 | - | 4, 5 | 255 | ||
AK002220.1 | - | 5, 6 | 522 | |||
AK008515.1 | - | 5, 6 | 512 | |||
AK009263.1 | - | 4, 5 | 650 | |||
AK009745.1 | - | 5, 6 | 504 | |||
AK010323.1 | - | 6, 7 | 548 | |||
AK129014.1 | - | 6, 7 | 3334 | |||
AK166519.1 | - | 6, 7 | 576 | |||
BC026765.1 | - | 6, 7 | 555 | 4166287 |

More Information
Other Species Targeted

