siRNA Details
Gene Symbol: 1700061I17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038029.1 | 1 | 139 | |||
GenBank mRNA | AK006854.1 | - | 1 | 141 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706840.1 | - | 1 | 141 | Chr. 3 - 117066933 - 117077765 [-] on Build GRCm38 |
