siRNA Details
Gene Symbol: Toporsos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045265.1 | 2 | 303 | |||
GenBank mRNA | AA614943.1 | - | 2 | 302 | ||
AK008077.1 | - | 301 | ||||
AK019894.1 | - | 2 | 303 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Toporsos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045265.1 | 2 | 303 | |||
GenBank mRNA | AA614943.1 | - | 2 | 302 | ||
AK008077.1 | - | 301 | ||||
AK019894.1 | - | 2 | 303 |