siRNA Details
Gene Symbol: 4930579G18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038053.1 | 6 | 575 | |||
GenBank mRNA | AK016319.1 | - | 6 | 576 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715958.1 | - | 6 | 576 | Chr. 14 - 54650511 - 54654576 [+] on Build GRCm38 |
