siRNA Details
Gene Symbol: BC006965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024085.1 | 1 | 230 | |||
GenBank mRNA | AK048256.1 | - | 1 | 231 | ||
AK145007.1 | - | 1 | 233 | |||
AK157980.1 | - | 1 | 237 | |||
BC006965.1 | - | 2 | 125 | |||
BC010247.1 | - | 2 | 122 |
