siRNA Details
Gene Symbol: 2900052L18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013668.1 | - | 2 | 537 | ||
AK034741.1 | - | 1 | 619 | |||
AK162948.1 | - | 1 | 720 |
Gene Symbol: LOC102640779 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094254.1 | - | 1 | 392 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712645.1 | - | 536 | Chr. - - [-] on Build |
