siRNA Details
Gene Symbol: 2810002D19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027831.1 | 2 | 649 | |||
GenBank mRNA | AK012638.1 | - | 2 | 650 | ||
AK016411.1 | - | 2 | 602 | |||
AK141492.1 | - | 569 | ||||
AK148028.1 | - | 601 | ||||
BC022909.1 | - | 1205 |
