siRNA Details
Gene Symbol: C130071C03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015561.2 | 4 | 392 | |||
GenBank mRNA | AK013308.1 | - | 2 | 298 | ||
AK039679.1 | - | 4 | 393 | |||
KF032589.1 | - | 341 | ||||
KF032590.1 | - | 263 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB633856.1 | - | 4 | 393 | Chr. 13 - 83728106 - 83734962 [+] on Build GRCm38 |
