siRNA Details
Gene Symbol: 4930579K19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029444.1 | 1 | 386 | |||
GenBank mRNA | AK016329.1 | - | 1 | 358 | ||
AK077474.1 | - | 1 | 368 | |||
AK087620.1 | - | 1 | 387 | |||
BC092140.1 | - | 1 | 327 |
