siRNA Details
Gene Symbol: Fcnaos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039020.1 | - | 4 | 890 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB180689.2 | - | 2 | 114 | Chr. 2 - 25625268 - 25631758 [+] on Build GRCm38 |
