siRNA Details
Gene Symbol: AI854703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027236.1 | 4 | 2086 | |||
GenBank mRNA | AK038960.1 | - | 4 | 2322 | ||
AK039299.1 | - | 2035 | ||||
AK129002.1 | - | 4 | 1703 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AI854703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027236.1 | 4 | 2086 | |||
GenBank mRNA | AK038960.1 | - | 4 | 2322 | ||
AK039299.1 | - | 2035 | ||||
AK129002.1 | - | 4 | 1703 |