siRNA Details
Gene Symbol: AI854703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027236.1 | 4 | 3055 | |||
GenBank mRNA | AK038960.1 | - | 4 | 3291 | ||
AK039299.1 | - | 3004 | ||||
AK085583.1 | - | 1 | 915 | |||
AK129002.1 | - | 4 | 2672 |
