siRNA Details
Gene Symbol: Gm12474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053462.1 | - | 3 | 405 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB665035.1 | - | 3 | 402 | Chr. 3 - 100489665 - 100523161 [+] on Build GRCm38 |
