siRNA Details
Gene Symbol: BC018473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003364.1 | 2 | 494 | |||
GenBank mRNA | AF118272.1 | - | 2 | 443 | ||
AF118273.1 | - | 2 | 492 | |||
AF118847.1 | - | 2 | 419 | |||
AK156340.1 | - | 2 | 332 | |||
BC018473.1 | - | 1 | 681 | |||
BY753224.1 | - | 2 | 337 |
