siRNA Details
Gene Symbol: Gm16294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046185.1 | 3 | 383 | |||
GenBank mRNA | AK034246.1 | - | 3 | 383 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB626236.1 | - | 3 | 383 | Chr. 15 - 40512136 - 40527277 [-] on Build GRCm38 |
