siRNA Details
Gene Symbol: 1700028I16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038042.1 | 3 | 556 | |||
GenBank mRNA | AK006461.1 | - | 3 | 554 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700028I16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038042.1 | 3 | 556 | |||
GenBank mRNA | AK006461.1 | - | 3 | 554 |