siRNA Details
Gene Symbol: Gm13939 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033473.1 | 5 | 635 | |||
GenBank mRNA | AK145809.1 | - | 5 | 636 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm13939 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033473.1 | 5 | 635 | |||
GenBank mRNA | AK145809.1 | - | 5 | 636 |