siRNA Details
Gene Symbol: Gm16551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045284.1 | 3 | 847 | |||
GenBank mRNA | AK051235.1 | - | 3 | 760 | ||
BC023444.1 | - | 3 | 719 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm16551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045284.1 | 3 | 847 | |||
GenBank mRNA | AK051235.1 | - | 3 | 760 | ||
BC023444.1 | - | 3 | 719 |