siRNA Details
Gene Symbol: Gm14164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033505.1 | 5 | 1975 | |||
GenBank mRNA | AY512911.1 | - | 5 | 1972 | ||
AY512942.1 | - | 5 | 1972 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm14164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033505.1 | 5 | 1975 | |||
GenBank mRNA | AY512911.1 | - | 5 | 1972 | ||
AY512942.1 | - | 5 | 1972 |