siRNA Details
Gene Symbol: A530040E14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087744.1 | - | 6 | 1306 | ||
AK142678.1 | - | 1812 | ||||
AK163084.1 | - | 3 | 1068 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_001037910.1 | - | 1068 | Chr. - - [-] on Build |
