siRNA Details
Gene Symbol: D6Ertd474e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027803.1 | 2 | 199 | |||
GenBank mRNA | AK139687.1 | - | 2 | 195 | ||
CJ059669.1 | - | 2 | 200 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: D6Ertd474e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027803.1 | 2 | 199 | |||
GenBank mRNA | AK139687.1 | - | 2 | 195 | ||
CJ059669.1 | - | 2 | 200 |