siRNA Details
Gene Symbol: Gm12298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033539.1 | 3, 4 | 542 | |||
GenBank mRNA | AK131836.1 | - | 3, 4 | 543 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707708.1 | - | 3, 4 | 543 | Chr. 11 - 66905721 - 66947086 [-] on Build GRCm38 |
