siRNA Details
Gene Symbol: Gm10433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045282.1 | 3 | 1066 | |||
GenBank mRNA | AK141725.1 | - | 3 | 1067 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DV662061.1 | - | 3 | 719 | Chr. 12 - 100188773 - 100192741 [+] on Build GRCm38 |
