siRNA Details
Gene Symbol: Gm10433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045282.1 | 3 | 1295 | |||
GenBank mRNA | AK141725.1 | - | 3 | 1296 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm10433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045282.1 | 3 | 1295 | |||
GenBank mRNA | AK141725.1 | - | 3 | 1296 |